4-Chlor-7-nitrobenzofurazan

Lieferant: CHEMPUR

KM06830.250 KM06830.1
CHMPKM06830.250EA 0 EUR
CHMPKM06830.250 MAYBKM06830.1
4-Chlor-7-nitrobenzofurazan
4-Chlor-7-nitrobenzofurazan
Formel: C₆H₂ClN₃O₃
MDL: MFCD00005808
CAS-Nummer: 10199-89-0
EINECS: 233-496-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR