3-(4-Methylphenyl)-1H-pyrazol-5-amin
Lieferant: CHEMPUR
Synonyme:
5-Amino-3-(4-methylphenyl)pyrazole, 3-Amino-5-(4-methylphenyl)-1H-pyrazole, 3-Amino-5-p-Tolylpyrazole, 5-(p-Tolyl)-1H-pyrazol-3-amine
KM06993.250
KM06993.1
CHMPKM06993.250EA
0
EUR
CHMPKM06993.250
MAYBKM06993.1
3-(4-Methylphenyl)-1H-pyrazol-5-amin
3-(4-Methylphenyl)-1H-pyrazol-5-amin
Formel:
C₁₀H₁₁N₃ Molecular Weight: 173,2 g/mol |
MDL:
MFCD00084887 CAS-Nummer: 78597-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...