3-Amino-3-(4-chlorphenyl)propionsäure

Lieferant: Thermo Scientific

KM08205.1 KM08205.10
MAYBKM08205.1EA 0 EUR
MAYBKM08205.1 MAYBKM08205.10
3-Amino-3-(4-chlorphenyl)propionsäure
3-Amino-3-(4-chlorphenyl)propionsäure
Formel: C₉H₁₀ClNO₂
MDL: MFCD00126370
CAS-Nummer: 19947-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR