tert-Butyl-3-aminobenzoat
Lieferant: Thermo Scientific
Synonyme:
tert-Butyl 3-aminobenzoate
KM08419.250
KM08419.1
MAYBKM08419.250EA
0
EUR
MAYBKM08419.250
MAYBKM08419.1
tert-Butyl-3-aminobenzoat
tert-Butyl-3-aminobenzoat
Formel:
C₁₁H₁₅NO₂ |
MDL:
MFCD00729048 CAS-Nummer: 92146-82-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...