1-[(2-Chlorethyl)sulfonyl]-4-fluorobenzol

Lieferant: Thermo Scientific

KM08544.50
MAYBKM08544.50EA 0 EUR
MAYBKM08544.50
1-[(2-Chlorethyl)sulfonyl]-4-fluorobenzol
1-[(2-Chlorethyl)sulfonyl]-4-fluorobenzol
Formel: C₈H₈ClFO₂S
MDL: MFCD00203483
CAS-Nummer: 33330-46-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR