N-(Chloracetyl)morpholin

Lieferant: Thermo Scientific

Synonyme: 4-(Chloroacetyl)morpholine, 4-(2-Chloroacetyl)morpholine

KM10055DA KM10055EA KM10055.50
MAYBKM10055DAEA 0 EUR
MAYBKM10055DA MAYBKM10055EA MAYBKM10055.50
N-(Chloracetyl)morpholin
N-(Chloracetyl)morpholin
Formel: C₆H₁₀ClNO₂
MDL: MFCD00721939
CAS-Nummer: 1440-61-5
EINECS: 215-880-3
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR