1,2-Dimethyl-4-imidazolsulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 1,2-Dimethyl-1H-imidazole-4-sulphonyl chloride

KM10104.0500
MAYBKM10104.0500EA 0 EUR
MAYBKM10104.0500
1,2-Dimethyl-4-imidazolsulfonylchlorid
1,2-Dimethyl-4-imidazolsulfonylchlorid
Formel: C₅H₇ClN₂O₂S
MDL: MFCD01313780
CAS-Nummer: 137049-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR