2-Hydroxy-6-methylisonicotinsäure

Lieferant: Thermo Scientific

Synonyme: 2-Hydroxy-6-methylpyridine-4-carboxylic acid

MO00027DA
MAYBMO00027DAEA 0 EUR
MAYBMO00027DA
2-Hydroxy-6-methylisonicotinsäure
2-Hydroxy-6-methylisonicotinsäure
Formel: C₇H₇NO₃
Molecular Weight: 153.15 g/mol
MDL: MFCD02682019
CAS-Nummer: 86454-13-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR