2,5-Dimethyl-3-thiophensulfonylchlorid
Lieferant: Thermo Scientific
MO00053DA
MAYBMO00053DAEA
0
EUR
MAYBMO00053DA
2,5-Dimethyl-3-thiophensulfonylchlorid
2,5-Dimethyl-3-thiophensulfonylchlorid
Formel:
C₆H₇ClO₂S₂ |
MDL:
MFCD03086201 CAS-Nummer: 97272-04-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...