5-Chlor-1,3-dimethyl-1H-pyrazol-4-carbonsäure

Lieferant: Thermo Scientific

MO00061CB MO00061DA
MAYBMO00061CBEA 0 EUR
MAYBMO00061CB MAYBMO00061DA
5-Chlor-1,3-dimethyl-1H-pyrazol-4-carbonsäure
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carboxylic acid
Formel: C₆H₇ClN₂O₂
MDL: MFCD00232043
CAS-Nummer: 27006-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR