(4-Nitrophenyl)methansulfonsäurechlorid

Lieferant: Thermo Scientific

Synonyme: 4-Nitro-α-toluenesulphonyl chloride

MO00918
MAYBMO00918EA 0 EUR
MAYBMO00918
(4-Nitrophenyl)methansulfonsäurechlorid
(4-Nitrophenyl)methanesulphonyl chloride
Formel: C₇H₆ClNO₄S
Molecular Weight: 235.64 g/mol
MDL: MFCD00034030
CAS-Nummer: 4025-75-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR