2-(Chlormethyl)benzonitril

Lieferant: Thermo Scientific

NRB01309.0250
MAYBNRB01309.0250EA 0 EUR
MAYBNRB01309.0250
2-(Chlormethyl)benzonitril
2-(Chlormethyl)benzonitril
Formel: C₈H₆ClN
Molecular Weight: 151,5929 g/mol
MDL: MFCD00019745
CAS-Nummer: 612-13-5
EINECS: 210-292-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR