3'-(Trifluormethyl)acetanilid

Lieferant: Thermo Scientific

Synonyme: N-(3-(Trifluormethyl)phenyl)acetamid, Ethansäure(3-(trifluormethyl)phenyl)amid, 3-Acetamidobenzotrifluorid

RDP00039.1 RDP00039.10 RDP00039.50
MAYBRDP00039.1EA 0 EUR
MAYBRDP00039.1 MAYBRDP00039.10 MAYBRDP00039.50
3'-(Trifluormethyl)acetanilid
3'-(Trifluormethyl)acetanilid
Formel: C₉H₈F₃NO
Molecular Weight: 203.16 g/mol
Siedepunkt: 287
Schmelzpunkt: 107…108 °C
MDL: MFCD00000383
CAS-Nummer: 351-36-0
EINECS: 206-512-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR