4-Iod-3,5-xylenol

Lieferant: Thermo Scientific

RDP00078.1 RDP00078.10 RDP00078.50
MAYBRDP00078.1EA 0 EUR
MAYBRDP00078.1 MAYBRDP00078.10 MAYBRDP00078.50
4-Iod-3,5-xylenol
4-Iod-3,5-xylenol
Formel: C₈H₉IO
MDL: MFCD00174284
CAS-Nummer: 80826-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR