4-Iod-o-toluidin

Lieferant: Thermo Scientific

Synonyme: 2-Amino-5-iodtoluol, 4-Iod-2-methylanilin

RDP00085.10 RDP00085.50
MAYBRDP00085.10EA 0 EUR
MAYBRDP00085.10 MAYBRDP00085.50
4-Iod-o-toluidin
4-Iod-o-toluidin
Formel: C₇H₈IN
Molecular Weight: 233.05 g/mol
Siedepunkt: 279
Schmelzpunkt: 91…92 °C
MDL: MFCD00025299
CAS-Nummer: 13194-68-8
EINECS: 236-154-2
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR