Ethyl-4-(trifluormethyl)benzoat

Lieferant: Thermo Scientific

Synonyme: Ethyl 4-(trifluoromethyl)benzoate

RDP00250.1
MAYBRDP00250.1EA 0 EUR
MAYBRDP00250.1
Ethyl-4-(trifluormethyl)benzoat
Ethyl-4-(trifluormethyl)benzoat
Formel: C₁₀H₉F₃O₂
Molecular Weight: 218,18 g/mol
MDL: MFCD00013563
CAS-Nummer: 583-02-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR