1,3-Bis(4-aminophenoxy)propan

Lieferant: Thermo Scientific

Synonyme: 4,4'-(Propane-1,3-diylbis(oxy))dianiline, 4,4'-(1,3-Propanediyl)dioxydianiline

RDR01946.0025
MAYBRDR01946.0025EA 0 EUR
MAYBRDR01946.0025
1,3-Bis(4-aminophenoxy)propan
1,3-Bis(4-aminophenoxy)propan
Formel: C₁₅H₁₈N₂O₂
Molecular Weight: 258,3200 g/mol
MDL: MFCD00025331
CAS-Nummer: 52980-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR