1,4-Bis(4-aminophenoxy)butan
Lieferant: Thermo Scientific
Synonyme:
4,4'-(Butane-1,4-diylbis(oxy))dianiline, 4,4'-(1,4-Butanediyl)dioxydianiline
RDR01947.0025
MAYBRDR01947.0025EA
0
EUR
MAYBRDR01947.0025
1,4-Bis(4-aminophenoxy)butan
1,4-Bis(4-aminophenoxy)butan
Formel:
C₁₆H₂₀N₂O₂ Molecular Weight: 272,3422 g/mol |
MDL:
MFCD00088690 CAS-Nummer: 6245-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...