N-(2,3-Dichlorphenyl)-2-chloracetamid

Lieferant: Thermo Scientific

RF00037.50
MAYBRF00037.50EA 0 EUR
MAYBRF00037.50
N-(2,3-Dichlorphenyl)-2-chloracetamid
N-(2,3-Dichlorphenyl)-2-chloracetamid
Formel: C₈H₆Cl₃NO
CAS-Nummer: 33560-47-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR