3-(2-Chlor-6-fluorphenyl)-5-methylisoxazol-4-carbonitril

Lieferant: Thermo Scientific

RF00744.1
MAYBRF00744.1EA 0 EUR
MAYBRF00744.1
3-(2-Chlor-6-fluorphenyl)-5-methylisoxazol-4-carbonitril
3-(2-Chlor-6-fluorphenyl)-5-methylisoxazol-4-carbonitril
Formel: C₁₁H₆ClFN₂O
MDL: MFCD00052551
CAS-Nummer: 175204-41-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR