Methyl-6-brom-2-chlorchinolin-4-carboxylat
Lieferant: Thermo Scientific
RF00961.1
MAYBRF00961.1EA
0
EUR
MAYBRF00961.1
Methyl-6-brom-2-chlorchinolin-4-carboxylat
Methyl-6-brom-2-chlorchinolin-4-carboxylat
Formel:
C₁₁H₇BrClNO₂ |
CAS-Nummer:
680213-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...