4-Iodphenylhydrazin

Lieferant: Thermo Scientific

Synonyme: (4-Iodophenyl)hydrazine

RF01374DA RF01374EA
MAYBRF01374DAEA 0 EUR
MAYBRF01374DA MAYBRF01374EA
4-Iodphenylhydrazin
4-Iodphenylhydrazin
Formel: C₆H₇IN₂
Molecular Weight: 234.04 g/mol
MDL: MFCD00052688
CAS-Nummer: 13116-27-3
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR