3-tert-Butylanilin

Lieferant: Thermo Scientific
Achtung

Synonyme: m-tert-Butylanilin

RF01575-0001
MAYBRF01575-0001EA 0 EUR
MAYBRF01575-0001
3-tert-Butylanilin
3-tert-Butylanilin
Formel: C₁₀H₁₅N
Molecular Weight: 149,24 g/mol
MDL: MFCD00125078
CAS-Nummer: 5369-19-7
EINECS: 226-361-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR