Benzamidoxim

Lieferant: Thermo Scientific

RF01827DA RF01827EA
MAYBRF01827DAEA 0 EUR
MAYBRF01827DA MAYBRF01827EA
Benzamidoxim
Benzamidoxim
Formel: C₇H₈N₂O
MDL: MFCD00031485
CAS-Nummer: 613-92-3
EINECS: 210-361-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR