4-(Trifluormethyl)benzamidoxim

Lieferant: Thermo Scientific

RF03217.250 RF03217DA
MAYBRF03217.250EA 0 EUR
MAYBRF03217.250 MAYBRF03217DA
4-(Trifluormethyl)benzamidoxim
4-(Trifluormethyl)benzamidoxim
Formel: C₈H₇F₃N₂O
Molecular Weight: 204,15 g/mol
MDL: MFCD00053042
CAS-Nummer: 22179-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR