Methyl-6-chlornicotinat

Lieferant: Thermo Scientific

Synonyme: Methyl 6-chloronicotinate

RF04027.1
MAYBRF04027.1EA 0 EUR
MAYBRF04027.1
Methyl-6-chlornicotinat
Methyl-6-chlornicotinat
Formel: C₇H₆ClNO₂
Molecular Weight: 171,58 g/mol
MDL: MFCD00023420
CAS-Nummer: 73781-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR