Propargylamin Hydrochlorid
Lieferant: Thermo Scientific
Synonyme:
2-Propinylammoniumchlorid, Propargylammoniumchlorid, Monopropargylamin Hydrochlorid, Prop-2-ynylamin Hydrochlorid, 3-Aminopropin Hydrochlorid
RH00060.1
RH00060.10
RH00060.50
MAYBRH00060.1EA
0
EUR
MAYBRH00060.1
MAYBRH00060.10
MAYBRH00060.50
Propargylamin Hydrochlorid
Propargylamin Hydrochlorid
Formel:
C₃H₆ClN Schmelzpunkt: 178…179 °C |
MDL:
MFCD00012907 CAS-Nummer: 15430-52-1 EINECS: 239-440-5 UN: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...