4-(Benzyloxy)-3-chlorbenzaldehyd
Lieferant: Thermo Scientific
RH00207.10
MAYBRH00207.10EA
0
EUR
MAYBRH00207.10
4-(Benzyloxy)-3-chlorbenzaldehyd
4-(Benzyloxy)-3-chlorbenzaldehyd
Formel:
C₁₄H₁₁ClO₂ Molecular Weight: 246,6900 g/mol |
MDL:
MFCD00831645 CAS-Nummer: 66422-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...