1-Acetyl-5-nitroindolin

Lieferant: Thermo Scientific

Synonyme: 1-(5-Nitroindolin-1-yl)ethanone

RH00434.1
MAYBRH00434.1EA 0 EUR
MAYBRH00434.1
1-Acetyl-5-nitroindolin
1-Acetyl-5-nitroindolin
Formel: C₁₀H₁₀N₂O₃
Molecular Weight: 206,2 g/mol
MDL: MFCD00005811
CAS-Nummer: 33632-27-8
EINECS: 251-609-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR