5-Bromindol
Lieferant: Thermo Scientific
Synonyme:
5-Bromo-1H-indole
RH00766.10
RH00766.50
MAYBRH00766.10EA
0
EUR
MAYBRH00766.10
MAYBRH00766.50
5-Bromindol
5-Bromindol
Formel:
C₈H₆BrN Schmelzpunkt: 90…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005670 CAS-Nummer: 10075-50-0 EINECS: 233-208-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...