2-(4-Methoxyphenoxy)-5-(trifluormethyl)anilin

Lieferant: Thermo Scientific

RH00953.1 RH00953.10
MAYBRH00953.1EA 0 EUR
MAYBRH00953.1 MAYBRH00953.10
2-(4-Methoxyphenoxy)-5-(trifluormethyl)anilin
2-(4-Methoxyphenoxy)-5-(trifluormethyl)anilin
Formel: C₁₄H₁₂F₃NO₂
MDL: MFCD00792432
CAS-Nummer: 62966-74-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR