2-Amino-4,6-dimethylpyrimidin

Lieferant: CHEMPUR

Synonyme: 4,6-Dimethylpyrimidin-2-ylamin, 4,6-Dimethylpyrimidin-2-amin

RJC01263.250 RJC01263.1
CHMPRJC01263.250EA 0 EUR
CHMPRJC01263.250 MAYBRJC01263.1
2-Amino-4,6-dimethylpyrimidin
2-Amino-4,6-dimethylpyrimidine
Formel: C₆H₉N₃
Molecular Weight: 123.16 g/mol
Schmelzpunkt: 152…155 °C
MDL: MFCD00006102
CAS-Nummer: 767-15-7
EINECS: 212-181-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR