2-Brom-5-nitrobenzoesäure
Lieferant: CHEMPUR
|
Achtung
|
RJC01300.250
RJC01300.1
CHMPRJC01300.250EA
0
EUR
CHMPRJC01300.250
MAYBRJC01300.1
2-Brom-5-nitrobenzoesäure
2-Bromo-5-nitrobenzoic acid
Formel:
C₇H₄BrNO₄ Molecular Weight: 246.02 g/mol Siedepunkt: 371 Schmelzpunkt: 178…180 °C |
MDL:
MFCD00134558 CAS-Nummer: 943-14-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...