2-Methyl-3,5-bis(trifluormethyl)anilin

Lieferant: Thermo Scientific

RJC02803-1
MAYBRJC02803-1EA 0 EUR
MAYBRJC02803-1
2-Methyl-3,5-bis(trifluormethyl)anilin
2-Methyl-3,5-bis(trifluormethyl)anilin
Formel: C₉H₇F₆N
CAS-Nummer: 243128-44-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR