4-(Dimethylamino)phenylisothiocyanat

Lieferant: Thermo Scientific

S05025.1 S05025.10 S05025.50
MAYBS05025.1EA 0 EUR
MAYBS05025.1 MAYBS05025.10 MAYBS05025.50
4-(Dimethylamino)phenylisothiocyanat
4-(Dimethylamino)phenylisothiocyanat
Formel: C₉H₁₀N₂S
MDL: MFCD00041093
CAS-Nummer: 2131-64-8
EINECS: 218-360-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR