trans-2,3-Dimethoxy-β-nitrostyrol

Lieferant: Thermo Scientific

Synonyme: 2,3-Dimethoxy-β-nitrostyrene

S14295.50
MAYBS14295.50EA 0 EUR
MAYBS14295.50
trans-2,3-Dimethoxy-β-nitrostyrol
trans-2,3-Dimethoxy-β-nitrostyrol
Formel: C₁₀H₁₁NO₄
Molecular Weight: 209.19 g/mol
MDL: MFCD00068082
CAS-Nummer: 37630-20-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR