5-Bromisatin

Lieferant: CHEMPUR

Synonyme: 5-Brom-2,3-indolindion, 5-Brom-1H-indol-2,3-dion

SB00153.250 SB00153.1
CHMPSB00153.250EA 0 EUR
CHMPSB00153.250 MAYBSB00153.1
5-Bromisatin
5-Bromisatin
Formel: C₈H₄BrNO₂
Molecular Weight: 226.03 g/mol
MDL: MFCD00149345
CAS-Nummer: 87-48-9
EINECS: 201-747-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR