2-Brom-5-methoxybenzoesäure

Lieferant: Thermo Scientific

SB00527.1 SB00527.10 SB00527.50
MAYBSB00527.1EA 0 EUR
MAYBSB00527.1 MAYBSB00527.10 MAYBSB00527.50
2-Brom-5-methoxybenzoesäure
2-Bromo-5-methoxybenzoic acid
Formel: C₈H₇BrO₃
MDL: MFCD00020214
CAS-Nummer: 22921-68-2
EINECS: 245-329-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR