2,4-Dichlor-5-methylbenzolsulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 2,4-Dichlor-5-methylbenzolsulfochlorid, 2,4-Dichlor-5-methylbenzolsulfonsäurechlorid, 2,4-Dichlor-5-methylbenzol-1-sulfonylchlorid

SB00644.1 SB00644.50
MAYBSB00644.1EA 0 EUR
MAYBSB00644.1 MAYBSB00644.50
2,4-Dichlor-5-methylbenzolsulfonylchlorid
2,4-Dichlor-5-methylbenzolsulfonylchlorid
Formel: C₇H₅Cl₃O₂S
Molecular Weight: 259.54
MDL: MFCD00044016
CAS-Nummer: 28286-86-4
EINECS: 248-938-1
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR