α-Chlor-m-xylol
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(Chloromethyl)-3-methylbenzene, 3-Methylbenzyl chloride
SB00682.1
SB00682.10
SB00682.50
MAYBSB00682.1EA
0
EUR
MAYBSB00682.1
MAYBSB00682.10
MAYBSB00682.50
α-Chlor-m-xylol
α-Chlor-m-xylol
Formel:
C₈H₉Cl Molecular Weight: 140.61 g/mol Siedepunkt: 196 Dichte: 1.064 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00000909 CAS-Nummer: 620-19-9 EINECS: 210-628-9 UN: 3265 ADR: 8 II Merck Index: 12,10224 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...