(4-Nitrophenyl)acetonitril
Lieferant: Thermo Scientific
Synonyme:
4-Nitrobenzylcyanid
SB00725.1
SB00725.10
SB00725.50
MAYBSB00725.1EA
0
EUR
MAYBSB00725.1
MAYBSB00725.10
MAYBSB00725.50
(4-Nitrophenyl)acetonitril
(4-Nitrophenyl)acetonitril
Formel:
C₈H₆N₂O₂ Molecular Weight: 162.15 g/mol Siedepunkt: 336 Schmelzpunkt: 108…113 °C |
MDL:
MFCD00007372 CAS-Nummer: 555-21-5 EINECS: 209-085-0 UN: 3439 ADR: 6.1 III Merck Index: 12,06688 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...