2-Chlor-5-nitropyridin
Lieferant: Thermo Scientific
|
Achtung
|
SB00747.1
SB00747.10
SB00747.50
MAYBSB00747.1EA
0
EUR
MAYBSB00747.1
MAYBSB00747.10
MAYBSB00747.50
2-Chlor-5-nitropyridin
2-Chloro-5-nitropyridine
Formel:
C₅H₃ClN₂O₂ Schmelzpunkt: 107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006240 CAS-Nummer: 4548-45-2 EINECS: 224-908-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...