1,4-Bis(2,2,2-trifluorethoxy)benzol

Lieferant: Thermo Scientific

SB00887.50
MAYBSB00887.50EA 0 EUR
MAYBSB00887.50
1,4-Bis(2,2,2-trifluorethoxy)benzol
1,4-Bis(2,2,2-trifluorethoxy)benzol
Formel: C₁₀H₈F₆O₂
Molecular Weight: 274,1597 g/mol
MDL: MFCD00042272
CAS-Nummer: 66300-61-6
EINECS: 266-304-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR