Methyl-2-(chlorsulfonyl)benzoat

Lieferant: Thermo Scientific

Synonyme: Methyl 2-(chlorosulphonyl)benzoate

SB00929.250
MAYBSB00929.250EA 0 EUR
MAYBSB00929.250
Methyl-2-(chlorsulfonyl)benzoat
Methyl-2-(chlorsulfonyl)benzoat
Formel: C₈H₇ClO₄S
Molecular Weight: 234,66 g/mol
MDL: MFCD00009797
CAS-Nummer: 26638-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR