(4-Nitrophenyl)essigsäure

Lieferant: Thermo Scientific

Synonyme: 4-Nitrophenylacetic acid

SB00940.1 SB00940.10 SB00940.50
MAYBSB00940.1EA 0 EUR
MAYBSB00940.1 MAYBSB00940.10 MAYBSB00940.50
(4-Nitrophenyl)essigsäure
(4-Nitrophenyl)essigsäure
Formel: (C₆H₄(NO₂))CH₂COOH
Schmelzpunkt: 150…153 °C
MDL: MFCD00007383
CAS-Nummer: 104-03-0
EINECS: 203-168-5
Merck Index: 12,06719

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR