Dimethyl-2-aminoterephthalat
Lieferant: Thermo Scientific
Synonyme:
Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate
SB01058.1
SB01058.10
SB01058.50
MAYBSB01058.1EA
0
EUR
MAYBSB01058.1
MAYBSB01058.10
MAYBSB01058.50
Dimethyl-2-aminoterephthalat
Dimethyl-2-aminoterephthalat
Formel:
C₁₀H₁₁NO₄ |
MDL:
MFCD00008427 CAS-Nummer: 5372-81-6 EINECS: 226-364-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...