Dimethyl-2-aminoterephthalat

Lieferant: Thermo Scientific

Synonyme: Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate

SB01058.1 SB01058.10 SB01058.50
MAYBSB01058.1EA 0 EUR
MAYBSB01058.1 MAYBSB01058.10 MAYBSB01058.50
Dimethyl-2-aminoterephthalat
Dimethyl-2-aminoterephthalat
Formel: C₁₀H₁₁NO₄
MDL: MFCD00008427
CAS-Nummer: 5372-81-6
EINECS: 226-364-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR