2-Isobutylthiazol

Lieferant: Thermo Scientific

SB01110.50
MAYBSB01110.50EA 0 EUR
MAYBSB01110.50
2-Isobutylthiazol
2-Isobutylthiazol
Formel: C₇H₁₁NS
Molecular Weight: 141,24 g/mol
Siedepunkt: 180 °C (1013 hPa)
Dichte: 0,9953 g/cm³ (25 °C)
MDL: MFCD00005334
CAS-Nummer: 18640-74-9
EINECS: 242-470-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR