2-Chlorbenzylalkohol
Lieferant: Thermo Scientific
Synonyme:
(2-Chlorophenyl)methanol
SB01129.1
SB01129.10
SB01129.50
MAYBSB01129.1EA
0
EUR
MAYBSB01129.1
MAYBSB01129.10
MAYBSB01129.50
2-Chlorbenzylalkohol
2-Chlorbenzylalkohol
Formel:
C₇H₇ClO Molecular Weight: 142.58 g/mol Siedepunkt: 227 Schmelzpunkt: 68…71 °C |
MDL:
MFCD00004604 CAS-Nummer: 17849-38-6 EINECS: 241-801-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...