4-Methoxyphenethylamin

Lieferant: Thermo Scientific

SB01162-0
MAYBSB01162-0EA 0 EUR
MAYBSB01162-0
4-Methoxyphenethylamin
4-Methoxyphenethylamin
Formel: C₉H₁₃NO
Molecular Weight: 151,21 g/mol
MDL: MFCD00008192
CAS-Nummer: 55-81-2
EINECS: 200-245-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR